Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01282188_m1
          See other RPGR GE Assays ›
          Gene Symbol:
          Rpgr
          Gene Name:
          retinitis pigmentosa GTPase regulator
          Gene Aliases:
          Rp3h
          Chromosome Location:
          Chr.X: 10158216 - 10216795 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01282188_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpgr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177950.1 NP_001171421.1 2 - 3 363 157
          NM_001177951.1 NP_001171422.1 2 - 3 363 157
          NM_001177952.1 NP_001171423.1 2 - 3 357 157
          NM_001177953.1 NP_001171424.1 2 - 3 363 157
          NM_001177954.1 NP_001171425.1 2 - 3 363 157
          NM_011285.2 NP_035415.1 2 - 3 363 157
          GenBank mRNA AF044677.1 - 2 - 3 281 157
          AJ238396.1 - 2 - 3 275 157
          AK017192.1 - 2 - 3 295 157
          AK029864.1 - 2 - 3 489 157
          AK038652.1 - 2 - 3 327 157
          AK081631.1 - 2 - 3 302 157
          AK136298.1 - 2 - 3 294 157
          AK138977.1 - 2 - 3 358 157
          BC012661.1 - 2 - 3 302 157

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003418.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK138977.1, NM_001177952.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpgr

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          cell projection organization
          intraciliary transport
          eye photoreceptor cell development
          response to stimulus
          retina morphogenesis in camera-type eye
          cilium assembly
          cellular response to light stimulus
          RNA binding
          guanyl-nucleotide exchange factor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00011801.mm.1 TC0X00003418.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011801.mm.1 NM_001177952.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011801.mm.1 NM_001177952.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 TC0X00003418.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177950.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177950.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177951.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177951.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177952.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177952.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_001177954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_011285.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011797.mm.1 NM_011285.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0