Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01282945_g1
          See other NAIP2 GE Assays ›
          Gene Symbol:
          Naip2
          Gene Name:
          NLR family, apoptosis inhibitory protein 2
          Gene Aliases:
          Birc1b, Naip-rs6
          Chromosome Location:
          Chr.13: 100144063 - 100202122 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01282945_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Naip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126182.2 NP_001119654.1 10 - 11 1307 65
          NM_010872.3 NP_035002.2 10 - 11 1378 65
          GenBank mRNA AF102871.1 - 1337 65
          AF135489.1 - 10 - 11 1348 65
          AF135490.1 - 10 - 11 1277 65
          AF381770.1 - 10 - 11 1277 65
          AY147000.1 - 10 - 11 1277 65
          AY147001.1 - 10 - 11 1277 65
          AY147002.1 - 8 - 9 1122 65
          AY147003.1 - 8 - 9 1165 65
          AY147004.1 - 9 - 10 1210 65
          AY147005.1 - 9 - 10 1212 65
          BC116626.1 - 9 - 10 1217 40086453 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002530.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Naip2

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          apoptotic process
          inflammatory response
          regulation of signal transduction
          detection of bacterium
          protein ubiquitination
          defense response to bacterium
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          innate immune response
          pyroptosis
          cellular response to estrogen stimulus
          inhibition of cysteine-type endopeptidase activity involved in apoptotic process
          nucleotide binding
          ubiquitin-protein transferase activity
          ATP binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300017077.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017077.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017077.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017081.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017081.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017081.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017080.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017080.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017080.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017088.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017088.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017088.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017082.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017082.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017082.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017074.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017074.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017074.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017078.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017078.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017078.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017087.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017087.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017075.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017075.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017075.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017076.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017076.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017076.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017079.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017079.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017079.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017083.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017083.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017083.mm.1 NM_010872.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017089.mm.1 TC1300002530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017089.mm.1 NM_001126182.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017089.mm.1 NM_010872.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0