Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081249.1 | NP_001074718.1 | 10 - 11 | 9808 | 111 | |
NM_001134474.1 | NP_001127946.1 | 9 - 10 | 4591 | 111 | ||
NM_001134475.1 | NP_001127947.1 | 8 - 9 | 1711 | 111 | ||
NM_019389.2 | NP_062262.2 | 9 - 10 | 6928 | 111 | ||
GenBank mRNA | AK045314.1 | - | 3 - 4 | 3263 | 111 | |
D16263.1 | - | 8 - 9 | 6500 | 111 | ||
D28599.1 | - | 10 - 11 | 9552 | 111 | ||
D32040.1 | - | 8 - 9 | 1443 | 111 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |