Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004149.1 | NP_001004149.1 | 1 - 2 | 41 | 64 | |
GenBank mRNA | BC070399.1 | - | 1 - 2 | 41 | 5694313 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |