Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300007160.mm.1 |
TC1300001128.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007160.mm.1 |
NM_001004149.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007160.mm.1 |
NM_001004149.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007159.mm.1 |
TC1300001128.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007159.mm.1 |
NM_001004149.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007159.mm.1 |
NM_001004149.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007158.mm.1 |
TC1300001128.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007158.mm.1 |
NM_001004149.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007158.mm.1 |
NM_001004149.1 |
Refseq |