Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01284163_m1
          See other PEX5L GE Assays ›
          Gene Symbol:
          Pex5l
          Gene Name:
          peroxisomal biogenesis factor 5-like
          Gene Aliases:
          1700016J08Rik, PXR2, Pex2, TRIP8b
          Chromosome Location:
          Chr.3: 32949631 - 33143273 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01284163_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pex5l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163516.2 NP_001156988.1 8 - 9 1227 78
          NM_001163517.2 NP_001156989.1 3 - 4 349 78
          NM_001289505.1 NP_001276434.1 10 - 11 1208 78
          NM_021483.4 NP_067458.2 10 - 11 1337 78
          GenBank mRNA AB032591.1 - 10 - 11 1332 78
          AK006035.1 - 3 - 4 350 78
          AK039764.1 - 10 - 11 1207 78
          AK083141.1 - 8 - 9 1119 78
          HQ646186.1 - 11 - 12 1556 78
          HQ646187.1 - 10 - 11 1490 78
          HQ646188.1 - 9 - 10 1385 78
          HQ646189.1 - 9 - 10 1418 78
          HQ646190.1 - 8 - 9 1313 78
          HQ646191.1 - 10 - 11 2146 78
          HQ646192.1 - 11 - 12 1560 78
          HQ646193.1 - 11 - 12 1466 78
          HQ646194.1 - 10 - 11 1361 78
          HQ646195.1 - 11 - 12 1334 78
          HQ646196.1 - 10 - 11 1229 78
          HQ646197.1 - 3 - 4 349 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001805.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pex5l

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          protein import into peroxisome matrix, docking
          regulation of membrane potential
          regulation of cAMP-mediated signaling
          regulated exocytosis
          maintenance of protein location
          positive regulation of corticotropin secretion
          cation transmembrane transport
          peroxisome targeting sequence binding
          peroxisome matrix targeting signal-1 binding
          intracellular cyclic nucleotide activated cation channel activity
          protein binding
          small GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300013997.mm.1 TC0300001805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300013997.mm.1 NM_001163516.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013997.mm.1 NM_001163517.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013997.mm.1 NM_001289505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013997.mm.1 NM_021483.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013998.mm.1 TC0300001805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300013998.mm.1 NM_001163516.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013998.mm.1 NM_001163517.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013998.mm.1 NM_001289505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013998.mm.1 NM_021483.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0