Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030119.1 | - | 15 - 16 | 2302 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | enhancer of rudimentary homolog pseudogene | 1700018B24Rik | -746.7kb | Anti-sense | Chr. 3 - 48605731 - 48609100 on NCBI Build GRCm38 | |
Downstream | protocadherin 18 | Pcdh18 |
3110038E07Rik, BB095589, PCDH68L |
381.4kb | Anti-sense | Chr. 3 - 49743291 - 49757382 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() |