Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01286698_m1
          See other SLC36A1OS GE Assays ›
          Gene Symbol:
          Slc36a1os
          Gene Name:
          solute carrier family 36 (proton/amino acid symporter), member 1, opposite strand
          Gene Aliases:
          4921508A21Rik
          Chromosome Location:
          Chr.11: 55191719 - 55197682 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01286698_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc36a1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046034.1 2 - 3 365 87
          GenBank mRNA AK014841.2 - 2 - 3 366 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714362.1 - 2 - 3 366 Chr. 11 - 55192917 - 55197682 [-] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 36 (proton/amino acid symporter),
          member 2
          Slc36a2 A530067G19Rik,
          PAT2,
          Tramd1
          -6.6kb Sense Chr. 11 - 55158468 - 55185095 on NCBI Build GRCm38
          Downstream solute carrier family 36 (proton/amino acid symporter),
          member 1
          Slc36a1 5830411H19Rik,
          AI839897,
          Pat1
          8.7kb Anti-sense Chr. 11 - 55204321 - 55236330 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002778.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2418536 AK014841.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158574 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2418536 AK014841.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158574 - -
          Whole Mouse Genome G4122A A_51_P442246 AK014841.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107708 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158574 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453696_PM_at - -
          Mouse Genome 430 2.0 Array 1453696_PM_at - -
          Mouse Genome 430 2.0 Array 1453696_at - -
          Affymetrix Mouse Clariom S Array PSR1100025223.mm.1 TC1100002778.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025223.mm.1 NR_046034.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025223.mm.1 NR_046034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025224.mm.1 TC1100002778.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025224.mm.1 NR_046034.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025224.mm.1 NR_046034.1 Refseq
          Mouse Expression Set 430 1453696_PM_at - -
          Mouse Expression Set 430 1453696_at - -
          Affymetrix HT Mouse Genome 430B Array 1453696_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453696_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 822257 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100540021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0