Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01287056_m1
          See other D830013O20RIK GE Assays ›
          Gene Symbol:
          D830013O20Rik
          Gene Name:
          RIKEN cDNA D830013O20 gene
          Gene Aliases:
          Gm891
          Chromosome Location:
          Chr.12: 73364075 - 73409563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01287056_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D830013O20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046013.1 2 - 3 311 100
          GenBank mRNA AK085818.1 - 2 - 3 306 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB661979.1 - 2 - 3 303 Chr. 12 - 73364120 - 73409555 [-] on Build GRCm38 100
          GenBankmRNA BB516053.2 - 2 - 3 283 Chr. 12 - 73364075 - 73409535 [-] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 38,
          member 6
          Slc38a6 AW322671,
          EG625098
          -9.4kb Anti-sense Chr. 12 - 73286794 - 73361998 on NCBI Build GRCm38
          Downstream predicted gene,
          34016
          Gm34016 93.4kb Sense Chr. 12 - 73489915 - 73509333 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001919.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK085818.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108462 AK085818.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134393 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10129903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09747989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09909152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09800485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10402699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10088494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10129902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10000808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09778270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10337239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09979333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09929045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10348747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09924482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10080305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10246635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10232234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10402687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09956523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09877561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10344516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09787462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10246626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09844967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09747990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10348750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10088492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09880483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09909159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10337241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10395764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09877562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10337236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09778262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10348757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09966344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09787459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28537149 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200013208.mm.1 TC1200001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013208.mm.1 NR_046013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013208.mm.1 NR_046013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013209.mm.1 TC1200001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013209.mm.1 NR_046013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013209.mm.1 NR_046013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013203.mm.1 TC1200001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013203.mm.1 NR_046013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013203.mm.1 NR_046013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013211.mm.1 TC1200001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013211.mm.1 NR_046013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013211.mm.1 NR_046013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013210.mm.1 TC1200001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013210.mm.1 NR_046013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013210.mm.1 NR_046013.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 694109 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0