Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01287122_m1
          See other RSAD2 GE Assays ›
          Gene Symbol:
          Rsad2
          Gene Name:
          radical S-adenosyl methionine domain containing 2
          Gene Aliases:
          2510004L01Rik, Vig1, cig5
          Chromosome Location:
          Chr.12: 26442743 - 26456478 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01287122_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021384.4 NP_067359.2 3 - 4 769 100
          GenBank mRNA AF236064.2 - 3 - 4 745 100
          AF442152.1 - 3 - 4 730 100
          AK010906.1 - 3 - 4 748 100
          AK036373.1 - 3 - 4 749 100
          AK149900.1 - 3 - 4 750 100
          AK150069.1 - 3 - 4 751 100
          AK150406.1 - 3 - 4 749 100
          AK150425.1 - 3 - 4 751 100
          AK150525.1 - 3 - 4 750 100
          AK150536.1 - 3 - 4 752 100
          AK150616.1 - 3 - 4 749 100
          AK150684.1 - 3 - 4 751 100
          AK150731.1 - 3 - 4 751 100
          AK150767.1 - 3 - 4 751 100
          AK150783.1 - 3 - 4 751 100
          AK150788.1 - 3 - 4 751 100
          AK150812.1 - 3 - 4 751 100
          AK150835.1 - 3 - 4 750 100
          AK151140.1 - 3 - 4 751 100
          AK151185.1 - 3 - 4 751 100
          AK151191.1 - 3 - 4 751 100
          AK151286.1 - 3 - 4 749 100
          AK151308.1 - 3 - 4 751 100
          AK151331.1 - 4 - 5 1186 100
          AK151339.1 - 3 - 4 749 100
          AK151389.1 - 3 - 4 751 100
          AK151410.1 - 3 - 4 750 100
          AK151412.1 - 3 - 4 749 100
          AK151414.1 - 3 - 4 750 100
          AK151476.1 - 3 - 4 751 100
          AK151502.1 - 3 - 4 751 100
          AK151538.1 - 3 - 4 751 100
          AK151770.1 - 3 - 4 749 100
          AK151825.1 - 3 - 4 749 100
          AK151846.1 - 3 - 4 749 100
          AK151906.1 - 3 - 4 751 100
          AK151909.1 - 3 - 4 749 100
          AK151940.1 - 3 - 4 749 100
          AK151954.1 - 3 - 4 751 100
          AK151971.1 - 3 - 4 751 100
          AK152050.1 - 3 - 4 749 100
          AK152065.1 - 3 - 4 751 100
          AK152075.1 - 3 - 4 751 100
          AK152235.1 - 3 - 4 749 100
          AK152239.1 - 3 - 4 751 100
          AK152241.1 - 3 - 4 751 100
          AK152310.1 - 3 - 4 749 100
          AK152354.1 - 3 - 4 749 100
          AK152493.1 - 3 - 4 751 100
          AK152600.1 - 3 - 4 749 100
          AK152601.1 - 3 - 4 751 100
          AK152631.1 - 3 - 4 749 100
          AK152644.1 - 3 - 4 751 100
          AK152664.1 - 3 - 4 751 100
          AK152701.1 - 3 - 4 100
          AK152796.1 - 3 - 4 749 100
          AK152861.1 - 3 - 4 749 100
          AK152880.1 - 3 - 4 749 100
          AK152895.1 - 3 - 4 751 100
          AK152934.1 - 3 - 4 751 100
          AK152962.1 - 3 - 4 749 100
          AK153003.1 - 3 - 4 751 100
          AK153024.1 - 3 - 4 751 100
          AK153062.1 - 3 - 4 753 100
          AK153077.1 - 3 - 4 749 100
          AK153078.1 - 3 - 4 749 100
          AK153232.1 - 3 - 4 751 100
          AK153234.1 - 3 - 4 751 100
          AK153306.1 - 3 - 4 749 100
          AK153318.1 - 3 - 4 751 100
          AK153324.1 - 3 - 4 751 100
          AK153350.1 - 4 - 5 1186 100
          AK153435.1 - 3 - 4 627 100
          AK153453.1 - 3 - 4 748 100
          AK153556.1 - 3 - 4 751 100
          AK159147.1 - 3 - 4 749 100
          BC057868.1 - 3 - 4 742 5327504 100
          CT010378.1 - 3 - 4 736 100

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK150069.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsad2

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          immune system process
          response to virus
          regulation of ossification
          positive regulation of toll-like receptor 7 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          CD4-positive, alpha-beta T cell activation
          CD4-positive, alpha-beta T cell differentiation
          negative regulation of viral genome replication
          innate immune response
          negative regulation of protein secretion
          defense response to virus
          positive regulation of T-helper 2 cell cytokine production
          catalytic activity
          protein binding
          protein self-association
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200010744.mm.1 TC1200001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010744.mm.1 NM_021384.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010744.mm.1 NM_021384.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010743.mm.1 TC1200001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010743.mm.1 NM_021384.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010743.mm.1 NM_021384.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010745.mm.1 TC1200001552.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010745.mm.1 NM_021384.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010745.mm.1 NM_021384.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0