Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01287538_g1
          See other 1700084C01RIK GE Assays ›
          Gene Symbol:
          1700084C01Rik
          Gene Name:
          RIKEN cDNA 1700084C01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 169928597 - 169935318 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01287538_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700084C01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033185.2 NP_001028357.1 3 - 4 421 100
          GenBank mRNA AK141447.1 - 3 - 4 422 100
          BC150846.1 - 2 - 3 346 9087757 100
          BC172001.1 - 2 - 3 349 9054287 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001563.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129107 NM_001033185.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129107 AK141447.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141449 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124602 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129107 NM_001033185.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129107 AK141447.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20819381 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04350583 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04350583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20819381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00944824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00557950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00073999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01052414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00719734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00719736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01074637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00511798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00086742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01074640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00067492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01016051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00511799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00719735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00511803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00745928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20819348 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351467 - -
          Affymetrix Mouse Gene 1.1 ST Array 10351465 - -
          Affymetrix Mouse Clariom S Array PSR0100012812.mm.1 TC0100001563.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012812.mm.1 NM_001033185.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012812.mm.1 NM_001033185.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012813.mm.1 TC0100001563.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012813.mm.1 NM_001033185.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012813.mm.1 NM_001033185.2 Refseq
          Murine Genome U74v2 Set 169871_PM_at - -
          Murine Genome U74v2 Set 169871_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 479600 - -
          ABI Mouse Genome Survey Microarray 757444 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229202 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862026 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862026 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0