Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288464_m1
          See other CD55 GE Assays ›
          Gene Symbol:
          Cd55
          Gene Name:
          CD55 antigen
          Gene Aliases:
          Daf, Daf-GPI, Daf1, GPI-DAF
          Chromosome Location:
          Chr.1: 130437196 - 130463012 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288464_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd55
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010016.2 NP_034146.2 9 - 10 1178 109
          GenBank mRNA AK160994.1 - 9 - 10 1156 109
          BC011314.1 - 9 - 10 1133 3663561 109
          CT010328.1 - 9 - 10 1107 109
          D63679.1 - 9 - 10 1089 109
          L41366.1 - 9 - 10 1122 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003037.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd55

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          complement activation, classical pathway
          regulation of complement activation, classical pathway
          negative regulation of catalytic activity
          innate immune response
          viral entry into host cell
          virus receptor activity
          enzyme inhibitor activity
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101585 AK160994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101585 NM_010016.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527752 AK005027.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527752 AK016197.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527752 AK128966.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527752 BC090620.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527752 NM_029231.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154139 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101585 AK160994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101585 NM_010016.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388412 CT010328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388412 D63679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388412 L41366.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388412 AK160994.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388412 BC011314.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P136303 AF143541.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199084 AK030285.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20610277 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119643 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04261928 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00184512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00184500 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00184496 - -
          Whole Mouse Genome G4122A A_51_P388412 CT010328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388412 D63679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388412 L41366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388412 AK160994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388412 BC011314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136303 AF143541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199084 AK030285.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00184496 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119643 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00184512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00064673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00064672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00919787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00064660 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00064659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20610314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01050471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00919791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00582818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00390830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00064664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00162836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00269376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00552106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00520023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00064672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00653024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00618115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00998355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00064673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00998354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00737872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00062917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00062897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00269390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00919786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00269397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00269395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20610220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357492 - -
          Affymetrix HT MG-430 PM Array Plate 1418762_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460242_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443906_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10357488 - -
          Affymetrix Mouse Gene 1.1 ST Array 10357480 - -
          Mouse Genome 430 2.0 Array 1443906_at - -
          Mouse Genome 430 2.0 Array 1443906_PM_at - -
          Mouse Genome 430 2.0 Array 1460242_PM_at - -
          Mouse Genome 430 2.0 Array 1418762_at - -
          Mouse Genome 430 2.0 Array 1418762_PM_at - -
          Mouse Genome 430 2.0 Array 1460242_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13136_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13136_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100024324.mm.1 TC0100003037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100024324.mm.1 NM_010016.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024325.mm.1 TC0100003037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100024325.mm.1 NM_010016.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024326.mm.1 TC0100003037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100024326.mm.1 NM_010016.3 Refseq
          Mouse Expression Set 430 1418762_PM_at - -
          Mouse Expression Set 430 1460242_at - -
          Mouse Expression Set 430 1418762_at - -
          Mouse Expression Set 430 1427632_PM_x_at - -
          Mouse Expression Set 430 1427632_x_at - -
          Mouse Expression Set 430 1460242_PM_at - -
          Mouse Expression Set 430 1443906_PM_at - -
          Mouse Expression Set 430 1443906_at - -
          Murine Genome U74v2 Set 103617_PM_at - -
          Murine Genome U74v2 Set 92198_PM_s_at - -
          Murine Genome U74v2 Set 103617_at - -
          Murine Genome U74v2 Set 92198_s_at - -
          Mouse Genome 430A 2.0 Array 1460242_at - -
          Mouse Genome 430A 2.0 Array 1427632_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1418762_PM_at - -
          Mouse Genome 430A 2.0 Array 1427632_x_at - -
          Mouse Genome 430A 2.0 Array 1460242_PM_at - -
          Mouse Genome 430A 2.0 Array 1418762_at - -
          Affymetrix HT Mouse Genome 430A Array 1418762_at - -
          Affymetrix HT Mouse Genome 430A Array 1418762_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460242_at - -
          Affymetrix HT Mouse Genome 430A Array 1460242_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443906_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443906_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 493734 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7040593 - -
          Illumina mouse-6 v1.0 expression beadchip 7040593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248714 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248714 - -
          mouse-6 v1.1 (Illumina) scl013136.4_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013136.4_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0