Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288490_m1
          See other TNIP1 GE Assays ›
          Gene Symbol:
          Tnip1
          Gene Name:
          TNFAIP3 interacting protein 1
          Gene Aliases:
          ABIN, ABIN-1, ABIN1, AU018810, Naf1, Nef, VAN
          Chromosome Location:
          Chr.11: 54910787 - 54962958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288490_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199275.2 NP_001186204.1 4 - 5 577 131
          NM_001199276.2 NP_001186205.1 2 - 3 247 131
          NM_001271455.1 NP_001258384.1 3 - 4 300 131
          NM_001271456.1 NP_001258385.1 4 - 5 472 131
          NM_021327.4 NP_067302.2 3 - 4 419 131
          GenBank mRNA AJ242777.1 - 2 - 3 219 131
          AJ242778.1 - 3 - 4 391 131
          AK135338.1 - 3 - 4 398 131
          AK156131.1 - 3 - 4 271 131
          AK170841.1 - 2 - 3 225 131
          AK172890.1 - 205 131
          BC008186.1 - 3 - 4 336 3589311 131
          BC008665.1 - 4 - 5 443 3598934 131
          BY197901.1 - 2 - 3 247 131
          CJ175540.1 - 2 - 3 242 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002770.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnip1

          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          leukocyte cell-cell adhesion
          glycoprotein biosynthetic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription by RNA polymerase II
          positive regulation of inflammatory response
          negative regulation of ERK1 and ERK2 cascade
          modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
          thiol-dependent deubiquitinase
          protein binding
          polyubiquitin modification-dependent protein binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 TC1100002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 NM_001199275.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 NM_001199276.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 NM_001271455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 NM_001271456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025113.mm.1 NM_021327.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025118.mm.1 TC1100002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025118.mm.1 NM_001271455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025118.mm.1 NM_001271456.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0