Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026225.3 | NP_080501.2 | 5 - 6 | 560 | 66 | |
GenBank mRNA | AB093286.1 | - | 5 - 6 | 532 | 66 | |
AK016663.1 | - | 5 - 6 | 573 | 66 | ||
AK031149.1 | - | 5 - 6 | 554 | 66 | ||
AK045223.1 | - | 5 - 6 | 554 | 66 | ||
AK048986.1 | - | 5 - 6 | 547 | 66 | ||
AK153320.1 | - | 5 - 6 | 549 | 66 | ||
BC025427.1 | - | 5 - 6 | 566 | 2649589 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |