Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288604_m1
          See other GM12216 GE Assays ›
          Gene Symbol:
          Gm12216
          Gene Name:
          predicted gene 12216
          Gene Aliases:
          602-19-115, OTTMUSG00000005650
          Chromosome Location:
          Chr.11: 53785489 - 53859256 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288604_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm12216
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033332.1 3 - 4 282 64
          GenBank mRNA AK138811.1 - 3 - 4 283 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY724574.1 - 3 - 4 283 Chr. 11 - 53786005 - 53859256 [-] on Build GRCm38 64
          GenBankmRNA BY096280.1 - 3 - 4 221 Chr. 11 - 53786197 - 53859196 [-] on Build GRCm38 64
          GenBankmRNA BF466244.1 - 3 - 4 171 Chr. 11 - 53785921 - 53859144 [-] on Build GRCm38 64
          GenBankmRNA AI449349.1 - 3 - 4 170 Chr. 11 - 53786145 - 53859144 [-] on Build GRCm38 64
          GenBankmRNA AI463030.1 - 3 - 4 170 Chr. 11 - 53784291 - 53859144 [-] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream interferon regulatory factor 1 Irf1 AU020929,
          Irf-1
          -7.6kb Anti-sense Chr. 11 - 53770014 - 53778374 on NCBI Build GRCm38
          Downstream solute carrier family 22 (organic cation transporter),
          member 5
          Slc22a5 Lstpl,
          Octn2,
          jvs
          50.9kb Sense Chr. 11 - 53864542 - 53891799 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100628 AK039036.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155688 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100620 NR_033332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100620 AK138811.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100628 AK039036.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02327823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02327922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07346395 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07346250 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02327872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02327859 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07346341 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126833 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02327922 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07346250 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09034501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08976595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09684071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09583678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09410242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09333659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09520129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09175979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09378625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09520112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09684075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08976593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09565849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09730050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09547266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09648107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09265368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09259850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09030264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09378623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09455062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09569563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09372928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09321286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09565838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09725447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09684074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09520128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09137940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09137943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09565836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09730052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09520116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09431189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27806836 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 130776_PM_at - -
          Murine Genome U74v2 Set 130776_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0