Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288712_m1
          See other CDKL3 GE Assays ›
          Gene Symbol:
          Cdkl3
          Gene Name:
          cyclin-dependent kinase-like 3
          Gene Aliases:
          AI852479, B230379H01Rik
          Chromosome Location:
          Chr.11: 52000858 - 52089778 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288712_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdkl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166653.1 NP_001160125.1 3 - 4 565 117
          NM_001166654.1 NP_001160126.1 3 - 4 655 117
          NM_001166655.1 NP_001160127.1 3 - 4 658 117
          NM_001166656.1 NP_001160128.1 3 - 4 565 117
          NM_001166657.1 NP_001160129.1 3 - 4 655 117
          NM_153785.4 NP_722480.3 3 - 4 655 117
          GenBank mRNA AK045036.1 - 2 - 3 1014 117
          AK045356.1 - 3 - 4 555 117
          AK046394.1 - 3 - 4 656 117
          AK050990.1 - 3 - 4 655 117
          AK078804.1 - 3 - 4 547 117
          AK090045.1 - 3 - 4 441 117
          AK166428.1 - 3 - 4 497 117
          BC028871.1 - 3 - 4 527 4241862 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000585.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdkl3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          negative regulation of axon extension
          positive regulation of dendrite morphogenesis
          dendrite extension
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 TC1100000585.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166653.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166653.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166654.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166655.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166655.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166656.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166656.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_001166657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_153785.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004544.mm.1 NM_153785.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix