Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01288886_g1
          See other SQSTM1 GE Assays ›
          Gene Symbol:
          Sqstm1
          Gene Name:
          sequestosome 1
          Gene Aliases:
          A170, OSF-6, Osi, STAP, STONE14, p62
          Chromosome Location:
          Chr.11: 50200152 - 50210820 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01288886_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sqstm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290769.1 NP_001277698.1 6 - 7 1033 94
          NM_011018.3 NP_035148.1 6 - 7 1033 94
          GenBank mRNA BC006019.1 - 6 - 7 1030 3487289 94
          CX240486.1 - 1 - 2 106 94
          E08146.1 - 6 - 7 991 94
          U40930.1 - 6 - 7 1003 94
          U57413.1 - 6 - 7 995 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002664.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sqstm1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          immune system process
          ubiquitin-dependent protein catabolic process
          autophagy
          apoptotic process
          positive regulation of macroautophagy
          cell differentiation
          regulation of I-kappaB kinase/NF-kappaB signaling
          protein heterooligomerization
          positive regulation of mitophagy in response to mitochondrial depolarization
          regulation of nucleic acid-templated transcription
          transcription coregulator activity
          protein kinase C binding
          protein binding
          zinc ion binding
          protein kinase binding
          SH2 domain binding
          identical protein binding
          protein homodimerization activity
          ubiquitin binding
          metal ion binding
          K63-linked polyubiquitin modification-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787326 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787321 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787325 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787327 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069714 U57413.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069714 E08146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069714 U40930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069714 NM_011018.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103628 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112747 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069714 U57413.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069714 E08146.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069714 U40930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069714 NM_011018.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119077 U40930.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787323 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02321872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112747 - -
          Whole Mouse Genome G4122A A_51_P119077 U40930.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110606 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110544 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09162993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09162987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09155619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09162992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09162994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787326 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787321 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09465333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09161070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09726682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09181546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09084930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09561303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09181554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09394068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09155619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385573 - -
          Affymetrix HT MG-430 PM Array Plate 1450957_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10375704 - -
          Affymetrix Mouse Gene 1.1 ST Array 10385572 - -
          Mouse Genome 430 2.0 Array 1444021_at - -
          Mouse Genome 430 2.0 Array 1450957_a_at - -
          Mouse Genome 430 2.0 Array 1444021_PM_at - -
          Mouse Genome 430 2.0 Array 1450957_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18412_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18412_at - -
          Affymetrix Mouse Clariom S Array PSR1100024319.mm.1 TC1100002664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024319.mm.1 NM_011018.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024318.mm.1 TC1100002664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024318.mm.1 NM_011018.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024316.mm.1 TC1100002664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024316.mm.1 NM_011018.3 Refseq
          Mouse Expression Set 430 1450957_a_at - -
          Mouse Expression Set 430 1450957_PM_a_at - -
          Mouse Expression Set 430 1444021_at - -
          Mouse Expression Set 430 1444021_PM_at - -
          Murine Genome U74v2 Set 101995_PM_at - -
          Murine Genome U74v2 Set 101995_at - -
          Mouse Genome 430A 2.0 Array 1450957_a_at - -
          Mouse Genome 430A 2.0 Array 1450957_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450957_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450957_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1444021_at - -
          Affymetrix HT Mouse Genome 430B Array 1444021_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911548 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6550056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710705 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710705 - -
          mouse-6 v1.1 (Illumina) scl018412.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018412.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710705 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710705 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018412.1_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0