Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100009608.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009608.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009608.mm.1 |
NM_007923.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009607.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009607.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009607.mm.1 |
NM_007923.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009610.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009610.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009610.mm.1 |
NM_007923.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009609.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009609.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009609.mm.1 |
NM_007923.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009598.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009598.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009598.mm.1 |
NM_007923.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009600.mm.1 |
TC0100001181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009600.mm.1 |
NM_007923.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009600.mm.1 |
NM_007923.2 |
Refseq |