Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01290097_m1
          See other IVNS1ABP GE Assays ›
          Gene Symbol:
          Ivns1abp
          Gene Name:
          influenza virus NS1A binding protein
          Gene Aliases:
          1190004M08Rik, 1700126I16Rik, AA960440, HSPC068, ND1, NS-1, NS1-BP, Nd1-L, Nd1-S, mKIAA0850
          Chromosome Location:
          Chr.1: 151344308 - 151364445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01290097_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ivns1abp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039511.1 NP_001034600.1 2 - 3 485 133
          NM_001039512.1 NP_001034601.1 2 - 3 485 133
          NM_054102.2 NP_473443.2 2 - 3 485 133
          GenBank mRNA AB055737.1 - 2 - 3 482 132
          AB055738.1 - 2 - 3 482 132
          AK007291.1 - 2 - 3 435 133
          AK144619.1 - 2 - 3 464 133
          AK144835.1 - 2 - 3 466 133
          AK145845.1 - 2 - 3 436 133
          AK146001.1 - 2 - 3 466 133
          AK146494.1 - 2 - 3 438 133
          AK146645.1 - 2 - 3 436 133
          AL355706.1 - 2 - 3 485 133
          BC004092.1 - 1 - 2 185 3593827 133
          BC040250.1 - 2 - 3 477 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001351.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB055737.1, AB055738.1, AK007291.1, AK144835.1, AK146001.1, AK146494.1, AK146645.1, BC004092.1, BC040250.1, NM_001039511.1, NM_001039512.1, NM_054102.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ivns1abp

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of intrinsic apoptotic signaling pathway
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 TC0100001351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_001039511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_001039511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_001039512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_001039512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_054102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011182.mm.1 NM_054102.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 TC0100001351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_001039511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_001039511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_001039512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_001039512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_054102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011181.mm.1 NM_054102.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 TC0100001351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_001039511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_001039511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_001039512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_001039512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_054102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011180.mm.1 NM_054102.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 TC0100001351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_001039511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_001039511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_001039512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_001039512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_054102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011179.mm.1 NM_054102.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0