Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01290116_m1
          See other EPB4.1L5 GE Assays ›
          Gene Symbol:
          Epb4.1l5
          Gene Name:
          erythrocyte protein band 4.1 like 5
          Gene Aliases:
          1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
          Chromosome Location:
          Chr.1: 119545032 - 119649136 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01290116_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epb4.1l5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113416.1 NP_001106887.1 10 - 11 1028 101
          NM_145506.4 NP_663481.2 10 - 11 1028 101
          GenBank mRNA AK034676.1 - 10 - 11 950 101
          AK037024.1 - 10 - 11 984 101
          AK040354.1 - 10 - 11 996 101
          AK044959.1 - 10 - 11 966 101
          AK044996.1 - 10 - 11 974 101
          AK047523.1 - 10 - 11 955 101
          AK079301.1 - 10 - 11 979 101
          AK136125.1 - 10 - 11 983 101
          AK161141.1 - 10 - 11 979 101
          AK167449.1 - 10 - 11 1028 101
          AK173198.1 - 10 - 11 962 101
          BC003937.1 - 10 - 11 1013 3486543 101
          BC011476.1 - 10 - 11 962 4014009 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002950.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Epb4.1l5

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis involved in differentiation
          in utero embryonic development
          somitogenesis
          epithelial to mesenchymal transition
          neural plate morphogenesis
          positive regulation of cell-matrix adhesion
          epithelial cell morphogenesis
          apical constriction
          substrate-dependent cell migration, cell attachment to substrate
          ectoderm development
          endoderm development
          mesoderm development
          mesoderm migration involved in gastrulation
          unidimensional cell growth
          posttranscriptional regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          actomyosin structure organization
          negative regulation of protein binding
          positive regulation of protein binding
          somite rostral/caudal axis specification
          axial mesoderm development
          axial mesoderm morphogenesis
          paraxial mesoderm development
          embryonic foregut morphogenesis
          positive regulation of focal adhesion assembly
          regulation of establishment of protein localization
          left/right axis specification
          cellular response to transforming growth factor beta stimulus
          protein binding
          cytoskeletal protein binding
          protein domain specific binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100023712.mm.1 TC0100002950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023712.mm.1 NM_001113416.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023712.mm.1 NM_001113416.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023712.mm.1 NM_145506.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023712.mm.1 NM_145506.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0