Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01290558_m1
          See other ATP1B1 GE Assays ›
          Gene Symbol:
          Atp1b1
          Gene Name:
          ATPase, Na+/K+ transporting, beta 1 polypeptide
          Gene Aliases:
          Atp4b, Atpb, Atpb-1, NKbeta1
          Chromosome Location:
          Chr.1: 164437099 - 164458355 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01290558_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009721.6 NP_033851.1 1 - 2 598 94
          GenBank mRNA AK005071.1 - 1 - 2 245 94
          AK010677.1 - 1 - 2 599 94
          AK140862.1 - 1 - 2 245 94
          AK160405.1 - 1 - 2 244 94
          AK161891.1 - 1 - 2 249 94
          BC027319.1 - 1 - 2 237 2648940 94
          BC094070.1 - 1 - 2 236 4974089 94
          X16646.1 - 1 - 2 135 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003422.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1b1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          blastocyst development
          positive regulation of defense response to virus by host
          transport
          ion transport
          potassium ion transport
          sodium ion transport
          cellular calcium ion homeostasis
          cellular sodium ion homeostasis
          cell adhesion
          regulation of gene expression
          regulation of cardiac muscle contraction by calcium ion signaling
          metal ion transport
          cellular potassium ion homeostasis
          positive regulation of ATPase activity
          sodium ion export across plasma membrane
          protein stabilization
          relaxation of cardiac muscle
          cardiac muscle contraction
          protein localization to plasma membrane
          membrane repolarization
          positive regulation of mitophagy in response to mitochondrial depolarization
          positive regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transport
          positive regulation of sodium ion export across plasma membrane
          positive regulation of calcium:sodium antiporter activity
          positive regulation of potassium ion import across plasma membrane
          potassium ion import across plasma membrane
          ATPase activator activity
          P-type sodium:potassium-exchanging transporter activity
          protein binding
          protein C-terminus binding
          MHC class II protein complex binding
          ATPase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100028115.mm.1 TC0100003422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028115.mm.1 NM_009721.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028116.mm.1 TC0100003422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028116.mm.1 NM_009721.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028118.mm.1 TC0100003422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028118.mm.1 NM_009721.6 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 690808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0