Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153555.2 | NP_705783.1 | 1 - 2 | 156 | 109 | |
GenBank mRNA | AK146825.1 | - | 1 - 2 | 83 | 109 | |
AK164799.1 | - | 1 - 2 | 83 | 109 | ||
BB851975.1 | - | 1 - 2 | 157 | 109 | ||
BC023704.1 | - | 1 - 2 | 97 | 5345701 | 109 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |