Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01290829_g1
          See other ATP1A2 GE Assays ›
          Gene Symbol:
          Atp1a2
          Gene Name:
          ATPase, Na+/K+ transporting, alpha 2 polypeptide
          Gene Aliases:
          AW060654, Atpa-3, mKIAA0778
          Chromosome Location:
          Chr.1: 172271709 - 172298064 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01290829_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178405.3 NP_848492.1 14 - 15 2139 72
          GenBank mRNA AK129212.1 - 14 - 15 2052 72
          AK137602.1 - 14 - 15 2075 72
          AK147331.1 - 14 - 15 2071 72
          AK148775.1 - 14 - 15 2071 72
          AK161840.1 - 14 - 15 2071 72
          AK164509.1 - 14 - 15 2075 72
          AK168703.1 - 14 - 15 2071 72
          BC025807.1 - 3 - 4 367 72
          BC036127.1 - 14 - 15 2063 4955003 72
          BC041774.1 - 11 - 12 1789 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003566.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1a2

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          regulation of the force of heart contraction
          regulation of respiratory gaseous exchange by nervous system process
          transport
          ion transport
          potassium ion transport
          sodium ion transport
          cellular sodium ion homeostasis
          regulation of muscle contraction
          regulation of smooth muscle contraction
          regulation of striated muscle contraction
          regulation of blood pressure
          adult locomotory behavior
          visual learning
          regulation of vasoconstriction
          cellular potassium ion homeostasis
          response to nicotine
          sodium ion export across plasma membrane
          locomotion
          negative regulation of heart contraction
          negative regulation of striated muscle contraction
          negative regulation of cytosolic calcium ion concentration
          cellular response to mechanical stimulus
          cellular response to steroid hormone stimulus
          regulation of cardiac muscle cell contraction
          negative regulation of calcium ion transmembrane transport
          negative regulation of calcium:sodium antiporter activity
          response to glycoside
          potassium ion import across plasma membrane
          nucleotide binding
          P-type sodium:potassium-exchanging transporter activity
          protein binding
          ATP binding
          hydrolase activity
          ATPase-coupled cation transmembrane transporter activity
          metal ion binding
          chaperone binding
          steroid hormone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100029305.mm.1 TC0100003566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029305.mm.1 NM_178405.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029305.mm.1 NM_178405.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029307.mm.1 TC0100003566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029307.mm.1 NM_178405.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029307.mm.1 NM_178405.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029306.mm.1 TC0100003566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029306.mm.1 NM_178405.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029306.mm.1 NM_178405.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0