Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038592.1 | NP_001033681.1 | 2 - 3 | 253 | 99 | |
NM_001038593.1 | NP_001033682.1 | 2 - 3 | 156 | 99 | ||
NM_001038594.1 | NP_001033683.1 | 2 - 3 | 95 | 99 | ||
NM_023505.2 | NP_075994.2 | 2 - 3 | 132 | 99 | ||
GenBank mRNA | AF276918.1 | - | 2 - 3 | 99 | 99 | |
AF380337.1 | - | 2 - 3 | 196 | 99 | ||
AK005853.1 | - | 2 - 3 | 156 | 99 | ||
AK142654.1 | - | 2 - 3 | 96 | 99 | ||
BC132031.1 | - | 2 - 3 | 175 | 40130308 | 99 | |
BC132033.1 | - | 2 - 3 | 175 | 40130310 | 99 | |
BC145304.1 | - | 2 - 3 | 175 | 9053858 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |