Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291575_m1
          See other BC018473 GE Assays ›
          Gene Symbol:
          BC018473
          Gene Name:
          cDNA sequence BC018473
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 116752167 - 116757883 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291575_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC018473
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003364.1 1 - 2 415 126
          GenBank mRNA AF118272.1 - 1 - 2 364 126
          AF118273.1 - 1 - 2 413 126
          BY753224.1 - 1 - 2 258 126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI156335.1 - 604 Chr. - - [-] on Build 126
          GenBankmRNA BI558599.1 - 606 Chr. - - [-] on Build 126
          GenBankmRNA BI650198.1 - 583 Chr. - - [-] on Build 126
          GenBankmRNA BI651971.1 - 1 - 2 236 Chr. 11 - 116752342 - 116755646 [+] on Build GRCm38 125
          GenBankmRNA BI652084.1 - 611 Chr. - - [-] on Build 126
          GenBankmRNA BI526741.1 - 333 Chr. - - [-] on Build 126
          GenBankmRNA BI409722.1 - 319 Chr. - - [-] on Build 126
          GenBankmRNA BY203931.1 - 1 - 2 Chr. 11 - 116752269 - 116753643 [+] on Build GRCm38 127
          GenBankmRNA BI650385.1 - 610 Chr. - - [-] on Build 126
          GenBankmRNA BI654663.1 - 605 Chr. - - [-] on Build 127
          GenBankmRNA BI555390.1 - 602 Chr. - - [-] on Build 126
          GenBankmRNA BI556263.1 - 1 - 2 130 Chr. 11 - 116752450 - 116755637 [+] on Build GRCm38 126
          GenBankmRNA BG862355.1 - 1 - 2 119 Chr. 11 - 116752477 - 116754995 [+] on Build GRCm38 126
          GenBankmRNA BI556615.1 - 596 Chr. - - [-] on Build 126
          GenBankmRNA BG974063.1 - 1 - 2 125 Chr. 11 - 116752465 - 116755592 [+] on Build GRCm38 126

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 St6galnac2 II,
          ST6GalNAc,
          Siat7,
          Siat7b
          -57.1kb Anti-sense Chr. 11 - 116676703 - 116695231 on NCBI Build GRCm38
          Downstream ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 St6galnac1 Siat7a 11.4kb Anti-sense Chr. 11 - 116765025 - 116775507 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001942.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100018462.mm.1 TC1100001942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018462.mm.1 NR_003364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018462.mm.1 NR_003364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018461.mm.1 TC1100001942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018461.mm.1 NR_003364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018461.mm.1 NR_003364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018465.mm.1 TC1100001942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018465.mm.1 NR_003364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018465.mm.1 NR_003364.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 723056 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0