Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291597_g1
          See other GM16880 GE Assays ›
          Gene Symbol:
          Gm16880
          Gene Name:
          predicted gene, 16880
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 136696211 - 136725728 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291597_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16880
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037986.1 3 - 4 395 75
          GenBank mRNA AK051055.1 - 3 - 4 395 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB655480.1 - 3 - 4 395 Chr. 1 - 136696211 - 136725170 [+] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          19705
          Gm19705 -33.8kb Sense Chr. 1 - 136683396 - 136690805 on NCBI Build GRCm38
          Downstream nuclear receptor subfamily 5,
          group A,
          member 2
          Nr5a2 AU020803,
          D1Ertd308e,
          Ftf,
          LRH-1,
          UF2-H3B
          117.7kb Anti-sense Chr. 1 - 136842562 - 136960448 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001276.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29085 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00195912 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116133 - -
          Whole Mouse Genome G4122A A_52_P545512 AK086995.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108819 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00243592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00552503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00552506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00837670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00837681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01014325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00243597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00023239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00584465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00837665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00443841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00597648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00419174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00584464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00443842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00988145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00638264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00988140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00475225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00504088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00210464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00552504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00023249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00514680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00210472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00893079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00988141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00419172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00616043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00030462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00337357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00864742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00616045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00837669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00030464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01004353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00837325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20647511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100010634.mm.1 TC0100001276.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100010634.mm.1 NR_037986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100010634.mm.1 NR_037986.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100010635.mm.1 TC0100001276.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100010635.mm.1 NR_037986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100010635.mm.1 NR_037986.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100010633.mm.1 TC0100001276.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100010633.mm.1 NR_037986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100010633.mm.1 NR_037986.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 431744 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0