Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291608_m1
          See other D830032E09RIK GE Assays ›
          Gene Symbol:
          D830032E09Rik
          Gene Name:
          RIKEN cDNA D830032E09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 107917259 - 107950947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291608_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D830032E09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102306.1 3 - 4 304 116
          NR_102307.1 4 - 5 418 116
          GenBank mRNA AK085905.1 - 3 - 4 305 116
          AK085943.1 - 4 - 5 393 116
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB662147.1 - 3 - 4 305 Chr. 1 - 107917259 - 107949469 [+] on Build GRCm38 116
          GenBankmRNA BB662215.1 - 4 - 5 393 Chr. 1 - 107917285 - 107949390 [+] on Build GRCm38 116

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream serine (or cysteine) peptidase inhibitor,
          clade B,
          member 8
          Serpinb8 CAP-2,
          CAP2,
          NK10,
          Spi8,
          ovalbumin
          -331.2kb Sense Chr. 1 - 107589925 - 107610472 on NCBI Build GRCm38
          Downstream cadherin 7,
          type 2
          Cdh7 9330156F07Rik,
          CDH7L1
          2.0mb Sense Chr. 1 - 109982327 - 110141917 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001034.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470494 AK004080.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470494 BC004035.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470494 BC018479.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470494 NM_026823.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140943 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04218977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04218942 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00154308 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04219008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04219045 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04218956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04218966 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04218907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04219062 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04219062 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00154308 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04218977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00828509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00157807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00655551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00706953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01107581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00931201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00129605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00453919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00706944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01066730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00453901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00172135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00002483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00998743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00931202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01129795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00167934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00077203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00002475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00794770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00453900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00655548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20515038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100008594.mm.1 TC0100001034.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008594.mm.1 NR_102306.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008594.mm.1 NR_102307.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0