Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291634_g1
          See other 4931419H13RIK GE Assays ›
          Gene Symbol:
          4931419H13Rik
          Gene Name:
          RIKEN cDNA 4931419H13 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 55055242 - 55084002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291634_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4931419H13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040593.1 7 - 8 834 80
          GenBank mRNA AK016464.1 - 7 - 8 834 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cyclin A1 Ccna1 -28.4kb Anti-sense Chr. 3 - 55045469 - 55055330 on NCBI Build GRCm38
          Downstream spastic paraplegia 20,
          spartin (Troyer syndrome) homolog (human)
          Spg20 AI840044,
          C79168,
          SPARTIN,
          TAHCCP1,
          mKIAA0610
          28.1kb Sense Chr. 3 - 55112074 - 55137332 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000415.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P84347 AK016464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P84347 AK016464.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33648 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100111 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P84347 AK016464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P84347 AK016464.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04912657 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04912694 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04912676 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00646134 - -
          Whole Mouse Genome G4122A A_52_P84347 AK016464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P84347 AK016464.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123202 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100111 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04912694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03123837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03064839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02946683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02589471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22151359 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300002934.mm.1 TC0300000415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300002934.mm.1 NR_040593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300002934.mm.1 NR_040593.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300002936.mm.1 TC0300000415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300002936.mm.1 NR_040593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300002936.mm.1 NR_040593.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300002935.mm.1 TC0300000415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300002935.mm.1 NR_040593.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300002935.mm.1 NR_040593.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 657818 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103780156 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0