Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292245_g1
          See other CCNA2 GE Assays ›
          Gene Symbol:
          Ccna2
          Gene Name:
          cyclin A2
          Gene Aliases:
          AA408589, Ccn-1, Ccn1, Ccna, CycA2, Cyca
          Chromosome Location:
          Chr.3: 36564865 - 36571996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292245_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccna2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009828.2 NP_033958.2 7 - 8 1391 66
          GenBank mRNA AK044924.1 - 7 - 8 1394 66
          AK135861.1 - 7 - 8 1363 66
          AK204936.1 - 3 - 4 309 66
          BC052730.1 - 7 - 8 1339 6816226 66
          X75483.1 - 7 - 8 1339 66
          Z26580.1 - 7 - 8 1348 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001856.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccna2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          mitotic cell cycle
          cell cycle
          regulation of G2/M transition of mitotic cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of fibroblast proliferation
          cell division
          protein binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058860 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058855 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 Z26580.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 BC052730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 NM_009828.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 Z26580.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 BC052730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 NM_009828.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43342 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150923 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 Z26580.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 BC052730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 NM_009828.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 AK044924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 Z26580.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 BC052730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 NM_009828.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 Z26580.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 BC052730.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 NM_009828.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 Z26580.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 BC052730.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 NM_009828.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22058842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04874255 - -
          Whole Mouse Genome G4122A A_51_P481920 Z26580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 BC052730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 NM_009828.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 AK044924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 Z26580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 BC052730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 NM_009828.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481920 AK044924.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128819 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02711955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02711950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03098243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02711956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058860 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22058862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22058765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02711948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22058864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10497837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497832 - -
          Affymetrix HT MG-430 PM Array Plate 1417910_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417911_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10497831 - -
          Mouse Genome 430 2.0 Array 1417910_PM_at - -
          Mouse Genome 430 2.0 Array 1417911_PM_at - -
          Mouse Genome 430 2.0 Array 1417910_at - -
          Mouse Genome 430 2.0 Array 1417911_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12428_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12428_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300014352.mm.1 TC0300001856.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014352.mm.1 NM_009828.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014352.mm.1 NM_009828.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014353.mm.1 TC0300001856.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014353.mm.1 NM_009828.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014353.mm.1 NM_009828.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014354.mm.1 TC0300001856.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014354.mm.1 NM_009828.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014354.mm.1 NM_009828.2 Refseq
          Mouse Expression Set 430 1417910_PM_at - -
          Mouse Expression Set 430 1417911_at - -
          Mouse Expression Set 430 1417910_at - -
          Mouse Expression Set 430 1417911_PM_at - -
          Murine Genome U74v2 Set 99186_at - -
          Murine Genome U74v2 Set 99186_PM_at - -
          Mouse Genome 430A 2.0 Array 1417911_PM_at - -
          Mouse Genome 430A 2.0 Array 1417910_PM_at - -
          Mouse Genome 430A 2.0 Array 1417911_at - -
          Mouse Genome 430A 2.0 Array 1417910_at - -
          Affymetrix HT Mouse Genome 430A Array 1417910_at - -
          Affymetrix HT Mouse Genome 430A Array 1417911_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417911_at - -
          Affymetrix HT Mouse Genome 430A Array 1417910_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5290075 - -
          Illumina mouse-6 v1.0 expression beadchip 5290075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2672442 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2672442 - -
          mouse-6 v1.1 (Illumina) scl22236.8_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22236.8_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2672442 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2672442 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0