Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
TC1200000503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_001165253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_001165253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_001165254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_001165254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_146034.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003514.mm.1 |
NM_146034.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
TC1200000503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_001165253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_001165253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_001165254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_001165254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_146034.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003515.mm.1 |
NM_146034.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
TC1200000503.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_001165253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_001165253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_001165254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_001165254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_146034.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003513.mm.1 |
NM_146034.3 |
Refseq |