Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292962_g1
          See other GEMIN2 GE Assays ›
          Gene Symbol:
          Gemin2
          Gene Name:
          gem (nuclear organelle) associated protein 2
          Gene Aliases:
          1700012N19Rik, Sip1
          Chromosome Location:
          Chr.12: 59013393 - 59027988 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292962_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gemin2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025656.4 NP_079932.2 9 - 10 853 104
          GenBank mRNA AK007515.1 - 9 - 10 854 104
          BC053424.1 - 9 - 10 1282 6336522 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000498.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gemin2

          Gene Ontology Categories:

          Function(s)

          spliceosomal complex assembly
          spliceosomal snRNP assembly
          mRNA splicing, via spliceosome
          mRNA processing
          RNA splicing

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469867 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469868 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107830 AK013414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107830 BC053424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107830 NM_025656.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107830 AK005928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107830 AK007515.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855196 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112599 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173861 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129267 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107830 AK013414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107830 BC053424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107830 NM_025656.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107830 AK005928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107830 AK007515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315101 AK013414.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315101 BC053424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315101 AK005928.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28469881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07635093 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129267 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07635079 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02526095 - -
          Whole Mouse Genome G4122A A_52_P544666 AK013414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544666 BC053424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544666 NM_025656.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544666 AK005928.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P544666 AK007515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315101 AK013414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315101 BC053424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315101 AK005928.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104526 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137446 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129267 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07635093 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10194052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10194043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10194050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10194049 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469868 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469867 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28469866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28469908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10160872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09748644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10104788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10194041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09748639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10041677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09784400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09855851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10045201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28469909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395894 - -
          Affymetrix HT MG-430 PM Array Plate 1451044_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458627_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395893 - -
          Mouse Genome 430 2.0 Array 1451044_PM_at - -
          Mouse Genome 430 2.0 Array 1451044_at - -
          Mouse Genome 430 2.0 Array 1458627_at - -
          Mouse Genome 430 2.0 Array 1458627_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66603_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66603_at - -
          Affymetrix Mouse Clariom S Array PSR1200003461.mm.1 TC1200000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003461.mm.1 NM_025656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003460.mm.1 TC1200000498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003460.mm.1 NM_025656.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003462.mm.1 TC1200000498.mm.1 MTA TC
          Mouse Expression Set 430 1451044_at - -
          Mouse Expression Set 430 1451044_PM_at - -
          Mouse Expression Set 430 1458627_at - -
          Mouse Expression Set 430 1458627_PM_at - -
          Murine Genome U74v2 Set 166221_PM_r_at - -
          Murine Genome U74v2 Set 166221_r_at - -
          Mouse Genome 430A 2.0 Array 1451044_PM_at - -
          Mouse Genome 430A 2.0 Array 1451044_at - -
          Affymetrix HT Mouse Genome 430A Array 1451044_at - -
          Affymetrix HT Mouse Genome 430A Array 1451044_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458627_at - -
          Affymetrix HT Mouse Genome 430B Array 1458627_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 414755 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2640706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255390 - -
          mouse-6 v1.1 (Illumina) scl43183.10.1_88-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43183.10.1_88-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2920800 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2920800 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255390 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc