Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295628_s1
          See other MAML3 GE Assays ›
          Gene Symbol:
          Maml3
          Gene Name:
          mastermind like 3 (Drosophila)
          Gene Aliases:
          AV234550, BC049812, Mam-2, mKIAA1816
          Chromosome Location:
          Chr.3: 51687320 - 52105085 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295628_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Maml3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004176.2 NP_001004176.2 2 2540 66
          GenBank mRNA AB553633.1 - 2 2106 66
          AK173269.1 - 1 750 66
          BC085318.1 - 1 429 30435466 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001960.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Maml3

          Gene Ontology Categories:

          Function(s) Process(es)

          Notch signaling pathway
          positive regulation of transcription by RNA polymerase II
          transcription coactivator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300014962.mm.1 TC0300001960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014962.mm.1 NM_001004176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014962.mm.1 NM_001004176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014963.mm.1 TC0300001960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014963.mm.1 NM_001004176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014963.mm.1 NM_001004176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014964.mm.1 TC0300001960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014964.mm.1 NM_001004176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014964.mm.1 NM_001004176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014960.mm.1 TC0300001960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014960.mm.1 NM_001004176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014960.mm.1 NM_001004176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014961.mm.1 TC0300001960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014961.mm.1 NM_001004176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014961.mm.1 NM_001004176.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc