Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295924_m1
          See other CCDC181 GE Assays ›
          Gene Symbol:
          Ccdc181
          Gene Name:
          coiled-coil domain containing 181
          Gene Aliases:
          4930455F23Rik, AI427889
          Chromosome Location:
          Chr.1: 164275598 - 164287847 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295924_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc181
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029115.3 NP_083391.2 4 - 5 1394 58
          GenBank mRNA AK149168.1 - 4 - 5 1389 58
          BC048086.1 - 4 - 5 1286 6390120 58
          BC060044.1 - 4 - 5 1393 58

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc181

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 BC060044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 AK076640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 BC048086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 NM_029115.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 BC060044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 AK076640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 BC048086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190437 NM_029115.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27364 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118740 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100673 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 BC060044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 AK076640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 BC048086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 NM_029115.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 BC060044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 AK076640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 BC048086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190437 NM_029115.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20788158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100673 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04337598 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118740 - -
          Whole Mouse Genome G4122A A_51_P190437 BC060044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 AK076640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 BC048086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 NM_029115.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 BC060044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 AK076640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 BC048086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190437 NM_029115.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101392 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118740 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00219386 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00219382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00219379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00075343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00219380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20788229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00223751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00922143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00461291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00922131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01124674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00219384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00922133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01091569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00333743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20788183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351270 - -
          Affymetrix HT MG-430 PM Array Plate 1435052_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351269 - -
          Mouse Genome 430 2.0 Array 1435052_at - -
          Mouse Genome 430 2.0 Array 1435052_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74895_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74895_PM_at - -
          Mouse Expression Set 430 1435052_at - -
          Mouse Expression Set 430 1435052_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435052_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435052_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 328824 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769153 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769153 - -
          mouse-6 v1.1 (Illumina) scl17282.7.92_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17282.7.92_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2908953 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2908953 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769153 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl17282.7.92_117-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0