Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01296089_m1
          See other NDUFAF2 GE Assays ›
          Gene Symbol:
          Ndufaf2
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2
          Gene Aliases:
          1810058I14Rik, C86051, Ndufa12l, mimitin
          Chromosome Location:
          Chr.13: 108052589 - 108158654 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01296089_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127346.1 NP_001120818.1 3 - 4 314 152
          GenBank mRNA AB183434.1 - 3 - 4 258 152
          AK007894.1 - 3 - 4 305 152
          AK166492.1 - 3 - 4 281 152
          AY262012.1 - 3 - 4 286 152
          BC116975.1 - 3 - 4 280 40126294 152
          BC116977.1 - 3 - 4 280 40126296 152
          BY251841.1 - 3 - 4 315 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002637.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory electron transport chain
          cellular respiration
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          reactive oxygen species metabolic process
          NADH dehydrogenase (ubiquinone) activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC116975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC116977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC144716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 AY262012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BY251841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC116975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC116977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BC144716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 AY262012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2947 BY251841.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751375 AK043350.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 152005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44113 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123770 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC116975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC116977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC144716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 AY262012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BY251841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC116975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC116977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BC144716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 AY262012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2947 BY251841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC116975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC116977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC144716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 AY262012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BY251841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC116975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC116977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BC144716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 AY262012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2947 BY251841.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10928147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10957834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10957837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10486736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10439524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10447441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11023527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10625294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10957833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10957835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10480406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11061543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10712944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10625299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11076640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10546787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11023523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29309689 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10412056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412057 - -
          Affymetrix HT MG-430 PM Array Plate 1452923_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10412054 - -
          Mouse Genome 430 2.0 Array 1452923_at - -
          Mouse Genome 430 2.0 Array 1452923_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75597_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75597_at - -
          Affymetrix Mouse Clariom S Array PSR1300017938.mm.1 TC1300002637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017938.mm.1 NM_001127346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017938.mm.1 NM_001127346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017935.mm.1 TC1300002637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017935.mm.1 NM_001127346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017935.mm.1 NM_001127346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017939.mm.1 TC1300002637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017939.mm.1 NM_001127346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017939.mm.1 NM_001127346.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017936.mm.1 TC1300002637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017936.mm.1 NM_001127346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017936.mm.1 NM_001127346.1 Refseq
          Mouse Expression Set 430 1452923_at - -
          Mouse Expression Set 430 1452923_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452923_at - -
          Affymetrix HT Mouse Genome 430B Array 1452923_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222174 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0