Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01297407_m1
          See other NOXRED1 GE Assays ›
          Gene Symbol:
          Noxred1
          Gene Name:
          NADP+ dependent oxidoreductase domain containing 1
          Gene Aliases:
          4933437F05Rik
          Chromosome Location:
          Chr.12: 87221123 - 87238774 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01297407_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Noxred1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027744.1 NP_082020.1 7 - 8 1096 87
          GenBank mRNA AK017096.1 - 7 - 8 1227 87
          BC125331.1 - 7 - 8 1096 40129846 87
          BC125333.1 - 7 - 8 1096 40129848 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002119.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proline biosynthetic process
          molecular_function
          pyrroline-5-carboxylate reductase activity
          N-ethylmaleimide reductase activity
          oxidoreductase activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 NM_027744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 BC125333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 NM_027744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170301 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119654 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 NM_027744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 AK017096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 BC125333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 NM_027744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119654 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02571116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170301 - -
          Whole Mouse Genome G4122A A_51_P505662 BC125331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 NM_027744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 AK017096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 BC125333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 NM_027744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505662 AK017096.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170301 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10377804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10377811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10243085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09933505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614903 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401746 - -
          Affymetrix HT MG-430 PM Array Plate 1430314_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401743 - -
          Mouse Genome 430 2.0 Array 1430314_PM_at - -
          Mouse Genome 430 2.0 Array 1430314_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71275_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71275_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200014692.mm.1 TC1200002119.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014692.mm.1 NM_027744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014692.mm.1 NM_027744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014691.mm.1 TC1200002119.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014691.mm.1 NM_027744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014691.mm.1 NM_027744.1 Refseq
          Mouse Expression Set 430 1430314_PM_at - -
          Mouse Expression Set 430 1430314_at - -
          Affymetrix HT Mouse Genome 430B Array 1430314_at - -
          Affymetrix HT Mouse Genome 430B Array 1430314_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 475429 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2630541 - -
          MouseWG-6 v2 Expression BeadChip 2120113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226545 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720236 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226545 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720236 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226545 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0