Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299400_m1
          See other DNAJC21 GE Assays ›
          Gene Symbol:
          Dnajc21
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 21
          Gene Aliases:
          4930461P20Rik, 9930116P15Rik
          Chromosome Location:
          Chr.15: 10446763 - 10470516 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299400_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030046.2 NP_084322.2 9 - 10 1338 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001201.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007165 AK141303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150389 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150389 AK140224.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2748 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 AK020688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 AK020688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150389 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150389 AK140224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007165 AK141303.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02972427 - -
          Whole Mouse Genome G4122A A_51_P439857 NM_030046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 AK020688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 NM_030046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11840635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11878832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11980289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11981071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12034088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12433953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12093895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12278104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11886250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10427713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427709 - -
          Affymetrix HT MG-430 PM Array Plate 1420221_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457281_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420219_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420220_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1460711_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10427704 - -
          Mouse Genome 430 2.0 Array 1420221_at - -
          Mouse Genome 430 2.0 Array 1420219_PM_at - -
          Mouse Genome 430 2.0 Array 1420220_PM_x_at - -
          Mouse Genome 430 2.0 Array 1457281_at - -
          Mouse Genome 430 2.0 Array 1457281_PM_at - -
          Mouse Genome 430 2.0 Array 1420221_PM_at - -
          Mouse Genome 430 2.0 Array 1460711_at - -
          Mouse Genome 430 2.0 Array 1460711_PM_at - -
          Mouse Genome 430 2.0 Array 1420219_at - -
          Mouse Genome 430 2.0 Array 1420220_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78244_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78244_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500009658.mm.1 TC1500001201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009658.mm.1 NM_030046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009658.mm.1 NM_030046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009656.mm.1 TC1500001201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009656.mm.1 NM_030046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009656.mm.1 NM_030046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009657.mm.1 TC1500001201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009657.mm.1 NM_030046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009657.mm.1 NM_030046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009660.mm.1 TC1500001201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009660.mm.1 NM_030046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009660.mm.1 NM_030046.2 Refseq
          Mouse Expression Set 430 1460711_PM_at - -
          Mouse Expression Set 430 1460711_at - -
          Mouse Expression Set 430 1420220_x_at - -
          Mouse Expression Set 430 1457281_PM_at - -
          Mouse Expression Set 430 1420219_PM_at - -
          Mouse Expression Set 430 1457281_at - -
          Mouse Expression Set 430 1420221_PM_at - -
          Mouse Expression Set 430 1420221_at - -
          Mouse Expression Set 430 1420219_at - -
          Mouse Expression Set 430 1420220_PM_x_at - -
          Murine Genome U74v2 Set 93478_PM_at - -
          Murine Genome U74v2 Set 93478_at - -
          Murine Genome U74v2 Set 169515_PM_r_at - -
          Murine Genome U74v2 Set 134385_PM_at - -
          Murine Genome U74v2 Set 169515_r_at - -
          Murine Genome U74v2 Set 134385_at - -
          Mouse Genome 430A 2.0 Array 1460711_at - -
          Mouse Genome 430A 2.0 Array 1460711_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460711_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460711_at - -
          Affymetrix HT Mouse Genome 430B Array 1420219_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457281_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1420219_at - -
          Affymetrix HT Mouse Genome 430B Array 1420221_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457281_at - -
          Affymetrix HT Mouse Genome 430B Array 1420221_at - -
          Affymetrix HT Mouse Genome 430B Array 1420220_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1420220_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678695 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0