Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299950_g1
          See other PDPK1 GE Assays ›
          Gene Symbol:
          Pdpk1
          Gene Name:
          3-phosphoinositide dependent protein kinase 1
          Gene Aliases:
          Pdk1
          Chromosome Location:
          Chr.17: 24073680 - 24150922 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299950_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdpk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080773.2 NP_001074242.1 3 - 4 463 112
          NM_001286662.1 NP_001273591.1 3 - 4 381 112
          NM_011062.4 NP_035192.2 3 - 4 463 112
          GenBank mRNA AF079535.1 - 3 - 4 420 112
          AF086625.1 - 3 - 4 429 112
          AF126294.1 - 3 - 4 404 112
          AK147149.1 - 3 - 4 455 112
          AK147835.1 - 3 - 4 438 112
          AK149162.1 - 3 - 4 396 112
          AK149289.1 - 3 - 4 421 112
          AK162677.1 - 3 - 4 423 112
          AK165564.1 - 3 - 4 400 112
          AY062008.1 - 3 - 4 359 112
          BC146001.1 - 3 - 4 400 40131304 112
          BC146003.1 - 3 - 4 400 40131306 112
          CJ046953.1 - 3 - 4 382 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001615.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdpk1

          Gene Ontology Categories:

          Function(s) Process(es)

          type B pancreatic cell development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of protein kinase activity
          hyperosmotic response
          signal transduction
          epidermal growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          regulation of endothelial cell migration
          negative regulation of cardiac muscle cell apoptotic process
          phosphorylation
          cell migration
          peptidyl-threonine phosphorylation
          calcium-mediated signaling
          negative regulation of transforming growth factor beta receptor signaling pathway
          activation of protein kinase B activity
          cellular response to insulin stimulus
          negative regulation of toll-like receptor signaling pathway
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of mast cell degranulation
          focal adhesion assembly
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to epidermal growth factor stimulus
          extrinsic apoptotic signaling pathway
          positive regulation of protein localization to plasma membrane
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          3-phosphoinositide-dependent protein kinase activity
          insulin receptor binding
          protein binding
          ATP binding
          phospholipase activator activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          phospholipase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700014188.mm.1 TC1700001615.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014188.mm.1 NM_001080773.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014188.mm.1 NM_001286662.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014188.mm.1 NM_011062.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014188.mm.1 NM_011062.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0