Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300828_m1
          See other A330009N23RIK GE Assays ›
          Gene Symbol:
          A330009N23Rik
          Gene Name:
          RIKEN cDNA A330009N23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 101221191 - 101225186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300828_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330009N23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045326.1 2 - 3 398 84
          NR_045327.1 2 - 3 319 84
          NR_045328.1 2 - 3 366 84
          GenBank mRNA AK038079.1 - 2 - 3 398 84
          AK039268.1 - 2 - 3 330 84
          BY254676.1 - 2 - 3 291 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB633145.1 - 2 - 3 330 Chr. 15 - 101221855 - 101225150 [-] on Build GRCm38 84
          GenBankmRNA BG088654.2 - 1 - 2 153 Chr. 15 - 101221728 - 101223633 [-] on Build GRCm38 84
          GenBankmRNA BU524515.1 - 378 Chr. - - [-] on Build 84
          GenBankmRNA BI220933.1 - 1 - 2 149 Chr. 15 - 101221659 - 101223629 [-] on Build GRCm38 84
          GenBankmRNA BY724193.1 - 2 - 3 366 Chr. 15 - 101221896 - 101225186 [-] on Build GRCm38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream activin A receptor,
          type 1B
          Acvr1b 6820432J04,
          ActR-IB,
          ActRIB,
          Acvrlk4,
          Alk4,
          SKR2
          -8.9kb Anti-sense Chr. 15 - 101174125 - 101212601 on NCBI Build GRCm38
          Downstream GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein Grasp tamalin 574.0 Anti-sense Chr. 15 - 101224207 - 101232756 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002256.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2278604 AK038079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2278604 BB164247.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2278604 BG076127.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156963 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2278604 AK038079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2278604 BB164247.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2278604 BG076127.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03116384 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123748 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03116384 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435844_PM_at - -
          Mouse Genome 430 2.0 Array 1435844_at - -
          Mouse Genome 430 2.0 Array 1435844_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500018528.mm.1 TC1500002256.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018528.mm.1 NR_045327.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018528.mm.1 NR_045327.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 TC1500002256.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045326.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045326.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045327.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045327.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045328.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018525.mm.1 NR_045328.1 Refseq
          Mouse Expression Set 430 1435844_PM_at - -
          Mouse Expression Set 430 1435844_at - -
          Affymetrix HT Mouse Genome 430B Array 1435844_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435844_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 699054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0