Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01301442_g1
          See other APPBP2OS GE Assays ›
          Gene Symbol:
          Appbp2os
          Gene Name:
          amyloid beta precursor protein (cytoplasmic tail) binding protein 2, opposite strand
          Gene Aliases:
          D630032N06Rik
          Chromosome Location:
          Chr.11: 85235166 - 85238304 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01301442_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Appbp2os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028329.1 2 - 3 276 153
          GenBank mRNA AK085477.1 - 2 - 3 253 153
          AK170617.1 - 2 - 3 277 153
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB501001.2 - 2 - 3 263 Chr. 11 - 85235188 - 85238300 [+] on Build GRCm38 153
          GenBankmRNA BB751395.1 - 1 - 2 92 Chr. 11 - 85237157 - 85238302 [+] on Build GRCm38 153
          GenBankmRNA BB661265.1 - 2 - 3 253 Chr. 11 - 85235189 - 85238304 [+] on Build GRCm38 153
          GenBankmRNA BY748530.1 - 2 - 3 272 Chr. 11 - 85235171 - 85238295 [+] on Build GRCm38 153

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream amyloid beta precursor protein (cytoplasmic tail) binding protein 2 Appbp2 1300003O07Rik,
          AI465480,
          PAT1
          -2.0kb Anti-sense Chr. 11 - 85191308 - 85235120 on NCBI Build GRCm38
          Downstream protein phosphatase 1D magnesium-dependent,
          delta isoform
          Ppm1d AV338790,
          Wip1
          73.7kb Sense Chr. 11 - 85311254 - 85347071 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001290.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 NR_028329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 AK085477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 AK170617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 BQ033447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 NR_028329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 AK085477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 AK170617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P400637 BQ033447.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129045 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170775 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 NR_028329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 AK085477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 AK170617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 BQ033447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 NR_028329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 AK085477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 AK170617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P400637 BQ033447.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170775 - -
          Whole Mouse Genome G4122A A_52_P67124 AK014613.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170775 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430265_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441641_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379889 - -
          Mouse Genome 430 2.0 Array 1441641_PM_at - -
          Mouse Genome 430 2.0 Array 1430265_at - -
          Mouse Genome 430 2.0 Array 1430265_PM_at - -
          Mouse Genome 430 2.0 Array 1441641_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 654810_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 654810_at - -
          Affymetrix Mouse Clariom S Array PSR1100012191.mm.1 TC1100001290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012191.mm.1 NR_028329.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012191.mm.1 NR_028329.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012192.mm.1 TC1100001290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012192.mm.1 NR_028329.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012192.mm.1 NR_028329.1 Refseq
          Mouse Expression Set 430 1430265_PM_at - -
          Mouse Expression Set 430 1430265_at - -
          Mouse Expression Set 430 1441641_PM_at - -
          Mouse Expression Set 430 1441641_at - -
          Mouse Genome 430A 2.0 Array 1430265_PM_at - -
          Mouse Genome 430A 2.0 Array 1430265_at - -
          Affymetrix HT Mouse Genome 430B Array 1441641_at - -
          Affymetrix HT Mouse Genome 430B Array 1441641_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436998 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104150035 - -
          Illumina mouse-6 v1.0 expression beadchip 104150035 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242074 - -
          Illumina Sentrix mouse-6.v1 bead array 104150035 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104150035 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0