Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01301513_g1
          See other BC003965 GE Assays ›
          Gene Symbol:
          BC003965
          Gene Name:
          cDNA sequence BC003965
          Gene Aliases:
          Ccsmst1
          Chromosome Location:
          Chr.17: 25184561 - 25187662 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01301513_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC003965
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183150.2 NP_898973.2 1 - 2 85 98
          GenBank mRNA BY262222.1 - 1 - 2 85 98
          CF729276.1 - 1 - 2 50 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000365.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_183150.2 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080748 BC094389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080748 NM_183150.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876814 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118534 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080748 BC094389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080748 NM_183150.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118534 - -
          Whole Mouse Genome G4122A A_51_P357838 BC094389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357838 NM_183150.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357838 AK141327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357838 BC003965.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101164 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13171995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13362279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442738 - -
          Affymetrix HT MG-430 PM Array Plate 1440466_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424545_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424546_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442736 - -
          Mouse Genome 430 2.0 Array 1424545_PM_at - -
          Mouse Genome 430 2.0 Array 1440466_at - -
          Mouse Genome 430 2.0 Array 1424546_at - -
          Mouse Genome 430 2.0 Array 1440466_PM_at - -
          Mouse Genome 430 2.0 Array 1424546_PM_at - -
          Mouse Genome 430 2.0 Array 1424545_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214489_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214489_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700003010.mm.1 TC1700000365.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003010.mm.1 NM_183150.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003010.mm.1 NM_183150.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003009.mm.1 TC1700000365.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003009.mm.1 NM_183150.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003009.mm.1 NM_183150.2 Refseq
          Mouse Expression Set 430 1424546_at - -
          Mouse Expression Set 430 1424545_PM_at - -
          Mouse Expression Set 430 1424546_PM_at - -
          Mouse Expression Set 430 1424545_at - -
          Mouse Expression Set 430 1440466_at - -
          Mouse Expression Set 430 1440466_PM_at - -
          Murine Genome U74v2 Set 166610_r_at - -
          Murine Genome U74v2 Set 169633_i_at - -
          Murine Genome U74v2 Set 168471_at - -
          Murine Genome U74v2 Set 168471_PM_at - -
          Murine Genome U74v2 Set 166610_PM_r_at - -
          Murine Genome U74v2 Set 169633_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1424546_PM_at - -
          Mouse Genome 430A 2.0 Array 1424546_at - -
          Mouse Genome 430A 2.0 Array 1424545_PM_at - -
          Mouse Genome 430A 2.0 Array 1424545_at - -
          Affymetrix HT Mouse Genome 430A Array 1424546_at - -
          Affymetrix HT Mouse Genome 430A Array 1424546_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424545_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424545_at - -
          Affymetrix HT Mouse Genome 430B Array 1440466_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440466_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256177 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline