Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302203_m1
          See other NUP62CL GE Assays ›
          Gene Symbol:
          Nup62cl
          Gene Name:
          nucleoporin 62 C-terminal like
          Gene Aliases:
          Gm719
          Chromosome Location:
          Chr.X: 140006802 - 140062707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302203_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup62cl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081668.1 NP_001075137.1 8 - 9 930 99
          GenBank mRNA BC100367.1 - 8 - 9 914 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003069.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976539 NM_001081668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976539 BC100367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962503 NM_001081668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962503 BC100367.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80866 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107606 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976539 NM_001081668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976539 BC100367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962503 NM_001081668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962503 BC100367.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P14518885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14527383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14724986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14804145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15136802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15173197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14576799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14712221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15145798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32696939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32697132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606983 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606979 - -
          Affymetrix Mouse Clariom S Array PSR0X00019278.mm.1 TC0X00003069.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019278.mm.1 NM_001081668.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019278.mm.1 NM_001081668.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 498001 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2532508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0