Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197024.1 | NP_001183953.1 | 2 - 3 | 57 | ||
NM_001290736.1 | NP_001277665.1 | 2 - 3 | 57 | |||
GenBank mRNA | BC059910.1 | - | 2 - 3 | 57 | ||
CJ068427.1 | - | 2 - 3 | 57 |
Set Membership: |
![]() ![]() ![]() ![]() |