Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302750_m1
          See other NUMB GE Assays ›
          Gene Symbol:
          Numb
          Gene Name:
          numb gene homolog (Drosophila)
          Gene Aliases:
          Nb
          Chromosome Location:
          Chr.12: 83794034 - 83921934 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302750_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Numb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136075.2 NP_001129547.1 1 - 2 148 83
          NM_001272055.1 NP_001258984.1 1 - 2 148 83
          NM_001272056.1 NP_001258985.1 3 - 4 378 83
          NM_010949.2 NP_035079.1 1 - 2 148 83
          NR_073563.1 4 - 5 495 83
          NR_073564.1 4 - 5 495 83
          GenBank mRNA AF169191.1 - 1 - 2 133 83
          AF169192.1 - 1 - 2 133 83
          AF170709.1 - 1 - 2 133 83
          AK004553.1 - 4 - 5 403 83
          BC033459.1 - 4 - 5 419 3991630 83
          BC050108.1 - 4 - 5 405 83
          U70674.1 - 3 - 4 299 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002053.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Numb

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          nervous system development
          neuroblast proliferation
          axonogenesis
          lateral ventricle development
          neuroblast division in subventricular zone
          positive regulation of cell migration
          positive regulation of polarized epithelial cell differentiation
          forebrain development
          adherens junction organization
          regulation of neuron differentiation
          negative regulation of Notch signaling pathway
          positive regulation of neurogenesis
          positive regulation of dendrite morphogenesis
          lung epithelial cell differentiation
          negative regulation of protein localization to plasma membrane
          protein binding
          beta-catenin binding
          alpha-catenin binding
          cadherin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 TC1200002053.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NM_001136075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NM_001272055.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NM_001272056.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NM_010949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NR_073563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014108.mm.1 NR_073564.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 TC1200002053.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NM_001136075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NM_001272055.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NM_001272056.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NM_010949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NR_073563.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014110.mm.1 NR_073564.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0