Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303211_m1
          See other MAPK14 GE Assays ›
          Gene Symbol:
          Mapk14
          Gene Name:
          mitogen-activated protein kinase 14
          Gene Aliases:
          CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha
          Chromosome Location:
          Chr.17: 28691342 - 28748405 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303211_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001168508.1 NP_001161980.1 8 - 9 1076 66
          GenBank mRNA AK089059.1 - 8 - 9 990 66
          BC085303.1 - 8 - 9 1052 5389457 66
          CX568915.1 - 7 - 8 470 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000484.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint signaling
          cell morphogenesis
          cartilage condensation
          angiogenesis
          placenta development
          chondrocyte differentiation
          glucose metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          transmembrane receptor protein serine/threonine kinase signaling pathway
          skeletal muscle tissue development
          positive regulation of gene expression
          positive regulation of myotube differentiation
          myoblast differentiation involved in skeletal muscle regeneration
          phosphorylation
          peptidyl-serine phosphorylation
          fatty acid oxidation
          osteoclast differentiation
          positive regulation of cyclase activity
          lipopolysaccharide-mediated signaling pathway
          response to muramyl dipeptide
          response to lipopolysaccharide
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          response to muscle stretch
          p38MAPK cascade
          positive regulation of protein import into nucleus
          signal transduction in response to DNA damage
          positive regulation of erythrocyte differentiation
          positive regulation of myoblast differentiation
          positive regulation of transcription by RNA polymerase II
          positive regulation of glucose import
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          striated muscle cell differentiation
          stress-activated MAPK cascade
          positive regulation of cardiac muscle cell proliferation
          cellular response to lipopolysaccharide
          cellular response to ionizing radiation
          negative regulation of canonical Wnt signaling pathway
          positive regulation of brown fat cell differentiation
          stress-induced premature senescence
          regulation of cytokine production involved in inflammatory response
          positive regulation of myoblast fusion
          positive regulation of reactive oxygen species metabolic process
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          NFAT protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700004209.mm.1 TC1700000484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004209.mm.1 NM_001168508.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004209.mm.1 NM_001168508.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix