Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303795_g1
          See other SMGC GE Assays ›
          Gene Symbol:
          Smgc
          Gene Name:
          submandibular gland protein C
          Gene Aliases:
          2310010P21Rik, DXImx49e, Muc19, Sfc21
          Chromosome Location:
          Chr.15: 91838328 - 91861435 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303795_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smgc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198927.3 NP_945121.1 17 - 18 2089 93
          GenBank mRNA AK132579.1 - 16 - 17 1992 93
          AY459348.1 - 17 - 18 2090 93
          BC022763.1 - 3 - 4 445 93
          BC049243.1 - 9 - 10 1094 93
          BC049976.1 - 3 - 4 463 93
          BC055489.1 - 3 - 4 435 93
          BC055490.1 - 7 - 8 895 93
          BC069958.1 - 8 - 9 921 93
          BC092073.1 - 8 - 9 1004 93
          BC138091.1 - 17 - 18 2090 8861110 93
          BC145136.1 - 12 - 13 1373 9053684 93
          EU089958.1 - 2 - 3 262 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000900.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smgc

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958720 BC069958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958720 BC092073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958720 BC049243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958720 BC049976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958720 BC055489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958720 BC069958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958720 BC092073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958720 BC049243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958720 BC049976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958720 BC055489.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30437787 - -
          Whole Mouse Genome G4122A A_51_P391560 BC069958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391560 BC092073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391560 BC049243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391560 BC049976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391560 BC055490.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12010967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30437815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12009896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11859095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11859092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12009908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12340770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12340750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30437813 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10426371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426375 - -
          Affymetrix HT MG-430 PM Array Plate 1428318_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431727_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10426370 - -
          Mouse Genome 430 2.0 Array 1428318_at - -
          Mouse Genome 430 2.0 Array 1431727_at - -
          Mouse Genome 430 2.0 Array 1428318_PM_at - -
          Mouse Genome 430 2.0 Array 1431727_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223809_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223809_at - -
          Affymetrix Mouse Clariom S Array PSR1500007186.mm.1 TC1500000900.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007186.mm.1 NM_198927.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007186.mm.1 NM_198927.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007185.mm.1 TC1500000900.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007185.mm.1 NM_198927.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007185.mm.1 NM_198927.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007184.mm.1 TC1500000900.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500007184.mm.1 NM_198927.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500007184.mm.1 NM_198927.3 Refseq
          Mouse Expression Set 430 1428318_PM_at - -
          Mouse Expression Set 430 1428318_at - -
          Mouse Expression Set 430 1431727_PM_at - -
          Mouse Expression Set 430 1431727_at - -
          Murine Genome U74v2 Set 131555_PM_at - -
          Murine Genome U74v2 Set 131555_at - -
          Murine Genome U74v2 Set 165493_r_at - -
          Murine Genome U74v2 Set 165493_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1431727_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431727_at - -
          Affymetrix HT Mouse Genome 430B Array 1428318_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428318_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980576 - -
          MouseWG-6 v2 Expression BeadChip 1780048 - -
          Illumina mouse-6 v1.0 expression beadchip 1980576 - -
          Illumina mouse-6 v1.0 expression beadchip 4540435 - -
          Illumina mouse-6 v1.0 expression beadchip 1780048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602496 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620853 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775962 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602496 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775962 - -
          mouse-6 v1.1 (Illumina) scl0002588.1_21-S - -
          mouse-6 v1.1 (Illumina) scl0002446.1_7-S - -
          mouse-6 v1.1 (Illumina) scl47600.16.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780048 - -
          Illumina Sentrix mouse-6.v1 bead array 1980576 - -
          Illumina Sentrix mouse-6.v1 bead array 4540435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002446.1_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002588.1_21-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47600.16.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780048 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980576 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620853 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2775962 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2602496 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2602496 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775962 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0