Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01305040_m1
          See other NPC2 GE Assays ›
          Gene Symbol:
          Npc2
          Gene Name:
          Niemann-Pick type C2
          Gene Aliases:
          2700012J19Rik, AA408070, AU045843, HE1
          Chromosome Location:
          Chr.12: 84754559 - 84773112 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01305040_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023409.4 NP_075898.1 1 - 2 328 67
          GenBank mRNA AB021289.1 - 1 - 2 83 67
          AI391386.1 - 1 - 2 328 67
          AK008603.1 - 1 - 2 115 67
          AK009127.1 - 1 - 2 145 67
          AK149805.1 - 1 - 2 117 67
          AK151067.1 - 1 - 2 115 67
          AK151133.1 - 1 - 2 116 67
          AK152236.1 - 1 - 2 115 67
          AK153020.1 - 1 - 2 115 67
          AK153203.1 - 1 - 2 115 67
          AK154838.1 - 1 - 2 115 67
          AK158624.1 - 1 - 2 296 67
          AK159154.1 - 1 - 2 162 67
          AK165263.1 - 1 - 2 141 67
          AK167869.1 - 1 - 2 116 67
          AK170327.1 - 1 - 2 116 67
          BC003471.1 - 1 - 2 130 2649564 67
          BC007190.1 - 1 - 2 162 3592515 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002081.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npc2

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          steroid metabolic process
          cholesterol metabolic process
          cholesterol transport
          intracellular sterol transport
          intracellular cholesterol transport
          cholesterol efflux
          cholesterol homeostasis
          cholesterol binding
          enzyme binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200014324.mm.1 TC1200002081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014324.mm.1 NM_023409.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014324.mm.1 NM_023409.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014322.mm.1 TC1200002081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014322.mm.1 NM_023409.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014322.mm.1 NM_023409.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014323.mm.1 TC1200002081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014323.mm.1 NM_023409.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014323.mm.1 NM_023409.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014325.mm.1 TC1200002081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014325.mm.1 NM_023409.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014325.mm.1 NM_023409.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617527 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0