Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01305413_g1
          See other 4930435F18RIK GE Assays ›
          Gene Symbol:
          4930435F18Rik
          Gene Name:
          RIKEN cDNA 4930435F18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 116802095 - 116838178 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01305413_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930435F18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015323.1 - 2 - 3 1338 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ISL1 transcription factor,
          LIM/homeodomain
          Isl1 -526.1kb Sense Chr. 13 - 116298257 - 116309721 on NCBI Build GRCm38
          Downstream poly (ADP-ribose) polymerase family,
          member 8
          Parp8 2810430O08Rik,
          ARTD16,
          D13Ertd275e
          14.5kb Sense Chr. 13 - 116850842 - 117025913 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2359643 AK015323.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AF140599.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK004114.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK149259.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK152012.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK159571.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK182752.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 AK183716.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 BC027396.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 BC051473.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 BC056992.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 BC083057.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 NM_019712.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 XM_001477172.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904285 XM_001480943.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167417 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08009853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02786529 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08009831 - -
          Whole Mouse Genome G4122A A_51_P373134 AK015323.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132577 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02786529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10818318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29359661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431547_PM_at - -
          Mouse Genome 430 2.0 Array 1431547_PM_at - -
          Mouse Genome 430 2.0 Array 1431547_at - -
          Mouse Expression Set 430 1431547_PM_at - -
          Mouse Expression Set 430 1431547_at - -
          Affymetrix HT Mouse Genome 430B Array 1431547_at - -
          Affymetrix HT Mouse Genome 430B Array 1431547_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0