Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306604_m1
          See other GPBP1 GE Assays ›
          Gene Symbol:
          Gpbp1
          Gene Name:
          GC-rich promoter binding protein 1
          Gene Aliases:
          1700034P14Rik, AU019836, D230035M11Rik, Gpbp, mGPBP
          Chromosome Location:
          Chr.13: 111425680 - 111490123 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306604_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122963.1 NP_001116435.1 2 - 3 1274 78
          NM_028487.4 NP_082763.3 2 - 3 1274 78
          GenBank mRNA AK082704.1 - 2 - 3 1092 78
          AK135728.1 - 2 - 3 1204 79
          AK136894.1 - 2 - 3 1052 78
          AK145318.1 - 2 - 3 1263 78
          AK151283.1 - 1 - 2 495 78
          AK161928.1 - 2 - 3 1062 78
          AK163370.1 - 2 - 3 1122 78
          AK163390.1 - 2 - 3 1276 78
          AK169406.1 - 2 - 3 1069 78
          AY382529.1 - 2 - 3 1072 78
          BC016083.1 - 1 - 2 292 4501399 78
          BC067075.1 - 2 - 3 1048 30614988 78
          BC076635.1 - 2 - 3 1104 78
          BC094888.1 - 2 - 3 1067 30628044 78
          BC096442.1 - 2 - 3 1091 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002661.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK082704.1, AK135728.1, AK145318.1, AK161928.1, AK163370.1, AK163390.1, AK169406.1, AY382529.1, BC016083.1, BC067075.1, BC094888.1, NM_001122963.1, NM_028487.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpbp1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          DNA binding
          DNA-binding transcription factor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300018025.mm.1 TC1300002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018025.mm.1 NM_001122963.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018025.mm.1 NM_001122963.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018025.mm.1 NM_028487.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018025.mm.1 NM_028487.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018026.mm.1 TC1300002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018026.mm.1 NM_001122963.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018026.mm.1 NM_001122963.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018026.mm.1 NM_028487.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018026.mm.1 NM_028487.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline