Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306852_g1
          See other KAT2A GE Assays ›
          Gene Symbol:
          Kat2a
          Gene Name:
          K(lysine) acetyltransferase 2A
          Gene Aliases:
          1110051E14Rik, AW212720, Gcn5, Gcn5l2, mmGCN5
          Chromosome Location:
          Chr.11: 100704746 - 100712467 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306852_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kat2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038010.2 NP_001033099.1 5 - 6 929 101
          NM_020004.5 NP_064388.2 5 - 6 929 101
          GenBank mRNA AF254441.1 - 5 - 6 866 101
          AK137358.1 - 5 - 6 906 101
          AK145205.1 - 4 - 5 827 101
          AK147042.1 - 5 - 6 877 101
          AK150225.1 - 5 - 6 907 101
          AK154866.1 - 5 - 6 917 101
          AK155347.1 - 5 - 6 910 102
          AK155791.1 - 5 - 6 930 101
          AK155842.1 - 5 - 6 930 101
          AK158079.1 - 5 - 6 917 101
          BC011088.1 - 5 - 6 635 101
          BC051952.1 - 5 - 6 911 101
          BC060050.1 - 5 - 6 911 101
          BC063752.1 - 5 - 6 914 6333090 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003780.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK155347.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kat2a

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          somitogenesis
          neural tube closure
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nervous system development
          cell population proliferation
          histone acetylation
          histone deubiquitination
          telencephalon development
          metencephalon development
          midbrain development
          positive regulation of cell projection organization
          regulation of protein stability
          positive regulation of histone acetylation
          multicellular organism growth
          histone H3 acetylation
          histone H4-K12 acetylation
          histone H3-K14 acetylation
          intracellular distribution of mitochondria
          cardiac muscle cell differentiation
          alpha-tubulin acetylation
          regulation of autophagy of mitochondrion
          regulation of nucleic acid-templated transcription
          positive regulation of protein targeting to mitochondrion
          positive regulation of transcription regulatory region DNA binding
          chromatin binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          N-acetyltransferase activity
          transcription factor binding
          H3 histone acetyltransferase activity
          acetyltransferase activity
          transferase activity
          acyltransferase activity
          protein phosphatase binding
          histone deacetylase binding
          histone acetyltransferase activity (H4-K12 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100034832.mm.1 TC1100003780.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034832.mm.1 NM_001038010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034832.mm.1 NM_001038010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034832.mm.1 NM_020004.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034832.mm.1 NM_020004.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0