Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01308018_mH
          See other HCN1 GE Assays ›
          Gene Symbol:
          Hcn1
          Gene Name:
          hyperpolarization-activated, cyclic nucleotide-gated K+ 1
          Gene Aliases:
          Bcng1, C630013B14Rik, HAC2
          Chromosome Location:
          Chr.13: 117602320 - 117987414 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01308018_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hcn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010408.3 NP_034538.2 1 - 2 772 119
          GenBank mRNA AF028737.1 - 1 - 2 392 119
          AJ225123.1 - 1 - 2 612 119
          AK158322.1 - 1 - 2 773 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001330.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hcn1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of membrane depolarization
          transport
          ion transport
          potassium ion transport
          sodium ion transport
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          regulation of membrane potential
          apical protein localization
          negative regulation of action potential
          retinal cone cell development
          response to calcium ion
          transmembrane transport
          cellular response to cAMP
          potassium ion transmembrane transport
          regulation of membrane hyperpolarization
          positive regulation of cation channel activity
          nucleotide binding
          ion channel activity
          intracellular cAMP-activated cation channel activity
          voltage-gated ion channel activity
          voltage-gated sodium channel activity
          voltage-gated potassium channel activity
          potassium channel activity
          sodium channel activity
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          protein C-terminus binding
          cAMP binding
          identical protein binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300008341.mm.1 TC1300001330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008341.mm.1 NM_010408.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008341.mm.1 NM_010408.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008343.mm.1 TC1300001330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008343.mm.1 NM_010408.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008343.mm.1 NM_010408.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008342.mm.1 TC1300001330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008342.mm.1 NM_010408.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008342.mm.1 NM_010408.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0